Learn More
Medchemexpress LLC Ilaprazole (sodium) | 172152-50-0 | 99.8% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYB2145
Encompass_Preferred
Ilaprazole (IY-81149) sodium is an orally active proton pump inhibitor. It irreversibly inhibits H+/K+-ATPase in a dose-dependent manner and is used for the research of gastric ulcers. It is also a potent TOPK (T-lymphokine-activated killer cell-originated protein kinase) inhibitor.
  • Orally active proton pump inhibitor.
  • Irreversibly inhibits H+/K+-ATPase with an IC50 of 6 μM.
  • Used for research of gastric ulcers.
  • Potent TOPK inhibitor.
Catalog No. 50-004-14442