Learn More
CPC Scientific H-Ile-Thr-Gln-Val-Pro-Phe-Ser-Val-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific CANS005A
Encompass
Natural immunodominant peptide fragment (209-217) derived from the gp 100 melanoma-associated antigen that shows intermediate binding affinity with the HLA-A2 molecule. It perhaps may be useful in the development of novel cancer immunotherapies.ONE-LETTER SEQUENCE: ITQVPFSVMOLECULAR FORMULA: C42H67N9O12MOLECULAR WEIGHT:890.05STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [319927-23-6], RESEARCH AREA: CancerREFERENCES: S.A. Rosenberg et al., Nature Med., 4, 321 (1998)
Catalog No. 50-210-8907