Learn More
Medchemexpress LLC Ilginatinib hydrochloride (NS-018 hydrochloride) | 1239358-85-0 | 98.9% | 425.89 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY19631B10MM/1ML
Encompass_Preferred
Ilginatinib hydrochloride (NS-018 hydrochloride) is a highly active and orally bioavailable JAK2 inhibitor. It demonstrates significant selectivity for JAK2 over other JAK family members and inhibits Src-family kinases. This compound exhibits potent inhibitory activity against cell lines with specific JAK2 mutations, showing minimal cytotoxicity to other hematopoietic cell lines.
  • Highly active and orally bioavailable JAK2 inhibitor
  • Shows selectivity for JAK2 over JAK1, JAK3, and Tyk2
  • Inhibits Src-family kinases (SRC and FYN)
  • Weakly inhibits ABL and FLT3
  • Potent inhibitory activity against cell lines with JAK2V617F or MPLW515L mutations
  • Minimal cytotoxicity against hematopoietic cell lines lacking constitutively activated JAK2
  • For research use only
Catalog No. 50-004-01243