Learn More
Medchemexpress LLC Ilicic acid | 4586-68-9 | 5 MG

Supplier: Medchemexpress LLC HYN40155MG
Ilicic acid is a sesquiterpene lactone isolated from *Ambrosia camphorata*. It is intended for research use only.
- High purity: 98.48%
- Appearance: Solid, white to off-white
- Molecular formula: C15H24O3
- Molecular weight: 252.35
- Origin: Ambrosia camphorata
- Soluble in DMSO (100 mg/mL)
- Soluble in 10% DMSO, 90% (20% SBE-β-CD in Saline) (≥ 2.5 mg/mL)
- Soluble in 10% DMSO, 90% Corn Oil (≥ 2.5 mg/mL)
- Recommended storage for powder: -20°C for 3 years or 4°C for 2 years
- Recommended storage for in solvent: -80°C for 6 months or -20°C for 1 month
- Ships at room temperature (continental US)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.