Learn More
AdipoGen Ilimaquinone (100 µg)
SDP
Supplier:  AdipoGen AGCN20038C100
Encompass_Preferred
Chemical. CAS: 71678-03-0. Formula: C22H30O4. MW: 358.5. Isolated from Anthelia sp. Cytoplasmic microtubule inhibitor. Cell permeable, antimicrobial, anti-inflammatory, antimitotic and cytotoxic compound. Induces a complete and reversible breakdown/disruption of Golgi membranes into smaller vesicular structures. Blocks the association of the ADP-ribosylation factor and beta-COP to the Golgi membrane. Blocks protein transport to the plasma membrane and inhibits gap junctional communication. HIV-1 inhibitor. Blocks the cytotoxicity of ricin and diphtheria toxin. S-Adenosylhomocysteinase hydrolase inhibitor and cellular methylations inhibitor. DNA polymerase beta lyase activity inhibitor. Anti-cancer compound.
Catalog No. 50-205-3385