Learn More
Enzo Life Sciences Ilimaquinone (100µg). CAS: 71678-03-0
SDP
Supplier:  Enzo Life Sciences BMLG4350100
Encompass_Preferred
Displays antimicrobial, anti-HIV and anti-inflammatory properties. Induces complete and reversible breakdown of Golgi membranes. Inhibits the association of β-COP and ADP-ribosylation factor to Golgi membranes, but unlike BFA, does not induce retrograde transport of Golgi enzymes into ER or cause random fusion of successive Golgi cisternae. Interacts with methylation enzymes. Shows anticancer activity. Alternative name: 3-[(Decahydro-1beta,2beta,4alphabeta-trimethyl-5-methylene-1-naphthyl)methyl]-2-hydroxy-5-methoxybenzoquinone. Purity: ≥98% (TLC). Solubility: 25mg/ml soluble in DMSO or 100% ethanol. Long Term Storage: -20°C.
Catalog No. 50-201-0884