Learn More
Medchemexpress LLC ILK-IN-3 100mg | 6975-75-3 | 232.24 g/mol | C10H12N6O | 100 MG
SDP
Supplier:  Medchemexpress LLC HY115677100MG
Encompass_Preferred
ILK-IN-3 is an orally active small-molecule inhibitor of integrin-linked kinase (ILK) used in research to study ILK-related signaling and anticancer responses. It has demonstrated kinase inhibition in cell-based assays and has been reported to enhance the antitumor activity of docetaxel in orthotopic LCC6 models.
  • Orally active integrin-linked kinase inhibitor for research use.
  • Reported to improve docetaxel efficacy in LCC6 orthotopic tumor models.
  • Demonstrates kinase inhibition at 10 μM against DYRK1, GSK3α/β, and CDK5/p25.
  • High purity (99.62%), suitable for biochemical and cell-based assays.
  • Light yellow solid with good solubility in DMSO (~83.33 mg/mL) when sonicated.
  • Store powder at -20°C for long-term stability; follow recommended solvent storage conditions for solutions.
Catalog No. 50-003-07706