Learn More
Abcam IM 54 (2-(1H-Indol-3-yl)-3-pentylamino-maleimide), Necrosis inhibitor, 5MG
Supplier:  Abcam AB1452315MG
Encompass_Preferred
MW 325.4 Da, Purity >98%. Potent, selective necrosis inhibitor (IC₅₀ = 3 μM, H₂O₂ exposed HeLa cells). Reduces oxidative stress-induced necrosis without effecting anticancer drug-induced apoptosis. Inhibits oxidative stress-induced necrotic cell death in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5376