Learn More
Medchemexpress LLC IM-54 | 861891-50-1 | 99.3% | 325.40 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10835150MG
Encompass_Preferred
IM-54 is a selective inhibitor of oxidative stress-induced necrosis. It demonstrates potent inhibitory activity against H2O2-induced necrosis. IM-54 acts as a potential cardioprotective agent and a biological tool for investigating the molecular mechanisms of cell death.
  • Selective inhibitor of oxidative stress-induced necrosis.
  • Potent inhibitory activity against H2O2-induced necrosis.
  • Potential cardioprotective agent.
  • Biological tool for investigating the molecular mechanisms of cell death.
Catalog No. 50-003-46688