Learn More
Selleck Chemical LLC IM156 5mg
SDP
Supplier:  Selleck Chemical LLC S96045MG
Encompass_Preferred
IM156 (HL156A), a metformin derivative, is a potent activator ofAMPK that increases AMPK phosphorylation. IM156 blocksoxidative phosphorylation (OXPHOS) through the inhibition ofcomplex I and increasesapoptosis. IM156 ameliorates various types of fibrosis and inhibits tumors. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3475
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.