Learn More
Biotage IMAC PHYTIP COLUMNS 1ML VOL
SDP
Supplier:  Biotage PTR912003
Encompass
IMAC PhyTip columns (1 mL volume) containing 20 µL affinity media per column, MEA 2. Incl. buffer kit. 96 pack size
Catalog No. NC2152705