Learn More
Medchemexpress LLC iMAC2 hydrochloride | 335166-00-2 | 93.98% | 542.11 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1100731MG
Encompass_Preferred
iMAC2 hydrochloride is a potent MAC (mitochondrial apoptosis-induced channel) inhibitor, with an IC50 of 28 nM and a LD50 of 15000 nM. It shows an anti-apoptotic effect and blocks cytochrome c release. It is for research use only and not sold to patients.
  • Potent MAC (mitochondrial apoptosis-induced channel) inhibitor
  • IC50 of 28 nM
  • LD50 of 15000 nM
  • Shows anti-apoptotic effect
  • Blocks cytochrome c release
Catalog No. 50-003-44069