Learn More
Medchemexpress LLC HY-101529 5mg Medchemexpress, Imanixil CAS:75689-93-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1015295MG
Encompass_Preferred
Medchemexpress, HY-101529 5mg Imanixil CAS:75689-93-9 Imanixil (HOE-402 free base) is an inducer of the LDL receptor (LDLR). Imanixil (HOE-402 free base) is also a potent cholesterol-lowering compound, which inhibits very low density-lipoprotein (VLDL) production, and consequently attenuates atherosclerosis development. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0637