Learn More
Medchemexpress LLC Imatinib-d8 | 1092942-82-9 | C29H23D8N7O | 1 MG
SDP
Supplier:  Medchemexpress LLC HY15463S1MG
Encompass_Preferred
Imatinib-d8 is a deuterium-labeled variant of Imatinib (STI571), recognized as an orally bioavailable tyrosine kinase inhibitor. This compound is characterized by its selective inhibition of key kinase activities, specifically targeting BCR/ABL, v-Abl, PDGFR, and c-kit. It is exclusively designated for research applications and is not intended for human use.
  • Deuterium labeled compound
  • Orally bioavailable
  • Selectively inhibits BCR/ABL, v-Abl, PDGFR, and c-kit kinase activity
  • Can be utilized as a tracer in various studies
  • Serves as an internal standard for precise quantitative analysis via techniques such as NMR, GC-MS, or LC-MS
Catalog No. 50-003-76897