Learn More
Medchemexpress LLC Imatinib Mesylate | 220127-57-1 | 99.9% | C30H35N7O4S | 10 MM 1 ML
SDP
Supplier:  Medchemexpress LLC HY5094610MM/1ML
Encompass_Preferred
Imatinib Mesylate (STI571 Mesylate) is an orally active tyrosine kinase inhibitor. It inhibits c-Kit, Bcr-Abl, and PDGFR with an IC50 of 100 nM.
  • Orally active tyrosine kinase inhibitor.
  • Inhibits c-Kit, Bcr-Abl, and PDGFR with an IC50 of 100 nM.
  • Cited in over 115 publications.
  • Used in research for cancer, infection, neurological disease, and inflammation/immunology.
  • Targets include c-Kit, Bcr-Abl, PDGFR, and Autophagy.
  • Active in Protein Tyrosine Kinase/RTK and Autophagy pathways.
Catalog No. 50-003-58179