Learn More
Cayman Chemical ImazalIl-d5sulfate 10mg
SDP
Supplier:  Cayman Chemical 3926110 MG
Encompass_Preferred
An internal standard for the quantification of imazalil by GC- or LC-MS
Catalog No. 50-291-8517