Learn More
Medchemexpress LLC Imazalil sulfate | 58594-72-2 | 99.6% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB1134A10MM/1ML
Encompass_Preferred
Imazalil sulfate is a fungicide with oral activity that strongly activates mPXR but not mCAR in mouse liver. It is commonly used to protect various agricultural crops against fungal attack. Imazalil sulfate induces developmental abnormalities, gut microbiota dysbiosis, and hepatic metabolism disorder. This product is for research use only.
  • Fungicide with oral activity
  • Commonly used to protect agricultural crops against fungal attack
  • Induces developmental abnormalities, gut microbiota dysbiosis, and hepatic metabolism disorder
  • Suppressed IL-17 mRNAs in EL4 cells
  • Induces reporter gene expression in mPXR-expressed HepG2 cells
  • Increases hepatic Cyp3a11 mRNA levels in male C57BL/6N mice
Catalog No. 50-004-13098