Learn More
Medchemexpress LLC Imazamox | 114311-32-9 | MFCD03427427 | 99.8% | 305.33 g/mol | C15H19N3O4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10042750MG
Encompass_Preferred
Imazamox is an imidazolinone-class herbicide supplied as a research standard for use as an acetolactate synthase (ALS) inhibitor in plant biology, herbicide-resistance research, and analytical method development. The solid standard is characterized by documented molecular weight and purity and is provided with supporting documentation for laboratory use.
  • Inhibits acetolactate synthase (ALS) in plants.
  • Provided as a high-purity analytical standard with COA available.
  • Molecular weight 305.33 g/mol for precise analytical calculations.
  • Suitable for reference, formulation, and method development.
  • Small solid quantities convenient for laboratory testing and assays.
Catalog No. 50-002-14824