Learn More
Medchemexpress LLC HY-12826 5mg Medchemexpress, IMB-XH1 CAS:292057-76-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY128265MG
Encompass_Preferred
Medchemexpress, HY-12826 5mg IMB-XH1 CAS:292057-76-2 IMB-XH1 is an inhibitor of myeloid cell factor 1 (Mcl-1). IMB-XH1 is a non-competitive Delhi metallo-β-lactamase (NDM-1) inhibitor. The IC50s of IMB-XH1 against metallo-β-lactamases NDM-1, IMP-4, ImiS and L1 are 0.4637 μM, 3.980 μM, 0.2287 μM and 1.158 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1161