Learn More
Medchemexpress LLC Imd-0560 | 439144-66-8 | MFCD | 98.7% | 428.12 g·mol⁻¹ | C15H8BrF6NO2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10566110MG
Encompass_Preferred
IMD-0560 is a small-molecule inhibitor of IκB kinase β (IKKβ) developed for research applications. It modulates the NF-κB signaling pathway and has been used in preclinical studies investigating inflammation and cancer-related processes. The material is supplied as a characterized research reagent with supporting analytical documentation.
  • Selective inhibition of IκB kinase β suitable for pathway and target validation studies.
  • High purity for reproducible in vitro and cell-based assays.
  • Available as solid and as a solution formulation for flexible experimental use.
  • Provided with safety data sheet, certificate of analysis, and analytical reports.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-41565