Learn More
Selleck Chemical LLC Imeglimin S9925-5MG
SDP
Supplier:  Selleck Chemical LLC S99255MG
Encompass_Preferred
Imeglimin (EMD 387008 IMEG PXL 008 RVT-1501) Hydrochloride is a new class of oral glucose-lowering agents Imeglimin normalizes glucose tolerance and insulin sensitivity by preserving mitochondrial function from oxidative stress and favoring lipid oxidation in liver of HFHSD mice
Catalog No. 50-000-23950