Learn More
Medchemexpress LLC Imeglimin (hydrochloride) | 775351-61-6 | 99.98% | 191.66 | 5 MG
Supplier: Medchemexpress LLC HY14771A5MG
Imeglimin hydrochloride is an oral glucose-lowering agent. It functions by reducing reactive oxygen species (ROS) production, increasing mitochondrial DNA, and improving mitochondrial function.
- Oral glucose-lowering agent.
- Reduces reactive oxygen species (ROS) production.
- Increases mitochondrial DNA.
- Improves mitochondrial function.
- Prevents human endothelial cell death by inhibiting mitochondrial permeability transition without inhibiting mitochondrial respiration.
- Significantly decreases hyperglycemia and restores normal glucose tolerance in models.