Learn More
Medchemexpress LLC Imetit dihydrobromide | 32385-58-3 | MFCD00153816 | ≥98.0% | 332.06 g/mol | C6H12Br2N4S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1011735MG
Encompass_Preferred
Imetit dihydrobromide is a potent, high-affinity histamine receptor agonist used in receptor pharmacology and cellular assays. It mimics histamine in functional assays and is supplied as a solid for in vitro research.
  • Potent H3 and H4 receptor agonist with Ki ≈ 0.3 nM (H3) and 2.7 nM (H4).
  • Demonstrated cellular activity (eosinophil shape change EC50 ≈ 25 nM).
  • High purity suitable for pharmacological studies (≥98% HPLC).
  • Molecular weight 332.06 g/mol and CAS 32385-58-3.
  • Available in small pack sizes for research use (1 mg, 5 mg, 10 mg).
Catalog No. 50-002-11662