Learn More
Hach Company Imhoff Settling Cone 1 L
Supplier:  Hach Company 206700
Encompass
Cone-Imhoff Settling-1 L-Polycarbonate-with Screw ClosureFully graduated cones for determination of small amounts of settleable matter in sewage by Imhoff Sedimentation Method Glass cone features reinforced tip for ruggedness while the Polycarbonate cone features a leak-proof removable closure for withdrawal of sediment for analysis Support ring support standand cone holders available and sold separately
Catalog No. 50-002-07199