Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Imidacloprid | 138261-41-3 | 98.5% | 1 ML
SDP

Supplier:  Medchemexpress LLC HYB083810MM/1ML

Encompass_Preferred

Imidacloprid is an effective neonicotinoid pesticide used for pest control in cereals, vegetables, tea, and cotton. It is intended for research use only and not for sale to patients.

  • Molecular weight: 255.66
  • Formula: C9H10ClN5O2
  • Appearance: Solid
  • Color: White to off-white
  • Solution storage: -80°C for 2 years or -20°C for 1 year
  • Shipping: Room temperature (continental US)

Catalog No. 50-004-07645


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.