Learn More
AdipoGen Imidacloprid
SDP
Supplier:  AdipoGen CDXI0121M100
Encompass_Preferred
Chemical. CAS 138261-41-3. Formula C9H10ClN5O2. MW 255.66. Imidacloprid is a broad-spectrum neonicotinoid insecticide that is effective against sucking and biting insects and insects of the Coleopteran and Lepidoptera orders. It is an antagonist of the post-synaptic nicotinic receptors in the insect central nervous system, blocking acetylcholine at the post-synaptic membrane and impairing normal nerve function. It is an agonist of insect acetylcholine receptors nAchRs with Ki values of 1.9, 2.1, 290, and 50 nM for M. persicae and D. melanogaster receptors, rat brain membranes and rat recombinant receptors containing alpha4beta2 subunits, respectively. In a mouse model of high-fat diet-induced obesity, imidacloprid increased body weight gain and adiposity and impairs glucose metabolism without increasing total food intake. Has been shown be to implicated in colony collapse disorder of honey bees. Can be used as a reference compound.
Catalog No. 50-256-0633