Learn More
Medchemexpress LLC Imidazo[1,2-a]pyridine-6-carboxamide, 2-(4-cyanophenyl)-N,N-bis(3-methylbutyl)-3-[3-(1-pyrrolidinyl)propyl]-, hydrochloride (1:1) | 1064662-40-3 | 99.5% | 550.18 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY11029
Encompass_Preferred
SNT-207707 is a selective, potent, and orally active melanocortin MC-4 receptor antagonist with an IC50 of 8 nM (binding) and 5 nM (function) on the MC-4 receptor. It can cross the blood-brain barrier. It is for research use only.
  • Selective, potent, and orally active melanocortin MC-4 receptor antagonist
  • Binds to the MC-4 receptor with an affinity of 8 nM
  • Shows more than 200-fold selectivity versus MC-3 and MC-5
  • Can cross the blood-brain barrier
  • Increases food intake in mice
  • Reduces tumor-induced weight loss
Catalog No. 50-003-45765