Learn More
Medchemexpress LLC Imidazo[5,1-f]triazin-4(1H)-one, 2-[2-ethoxy-5-(4-morpholinylsulfonyl)phenyl]-5-methyl | 224788-36-7 | MFCD01346226 | 99.6% | 461.53 g/mol | C21H27N5O5S | 5 MG

Supplier: Medchemexpress LLC HY1420595MG
PDE5-IN-4 is a small-molecule phosphodiesterase 5 (PDE5) inhibitor supplied for research use. The compound is provided as a solid with high reported purity and is used in preclinical studies investigating acute myocardial infarction and reperfusion injury, gastrointestinal disorders, diabetes-related tissue damage, and liver failure.
- High purity of 99.6%.
- Molecular formula C21H27N5O5S.
- Molecular weight 461.53 g/mol.
- Available in small pack sizes, including 5 mg.
- Powder storage: -20°C for up to 3 years or 4°C for up to 2 years.
- In solvent storage: -80°C for up to 6 months or -20°C for up to 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.