Learn More
TARGETMOL CHEMICALS INC Imidazole-5-propionic acid
SDP
Supplier:  TARGETMOL CHEMICALS INC T9494500MG
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Imidazole-5-propionic acid (Imidazolylpropionic acid) is a product of histidine metabolism and may involve oxidation or transamination. Imidazole-5-propionic acid is potent inhibitors of G. candidum histidinol dehydrogenase, showing IC50 values as low as 3.17 microM. Purity 98.43%
Catalog No. 50-288-0405