Learn More
Medchemexpress LLC Imidazole-5-propionic acid | 1074-59-5 | MFCD00237415 | 100.0% | 140.14 | C6H8N2O2 | 100 MG

Supplier: Medchemexpress LLC HYW045271100MG
Imidazole-5-propionic acid is a histidine metabolite supplied as a high-purity research standard intended for biochemical and metabolic studies. The compound is provided with a certificate of analysis and is for research use only.
- High purity (99.99%).
- Molecular weight 140.14 g/mol.
- Molecular formula C6H8N2O2.
- Available in multiple pack sizes, including 100 MG.
- Intended for research use only; not for clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.