Learn More
Medchemexpress LLC Imidazole-5-propionic acid | 1074-59-5 | MFCD00237415 | 100.0% | 140.14 g/mol | C6H8N2O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW04527110MG
Encompass_Preferred
Imidazole-5-propionic acid is an imidazole-containing metabolite of histidine used as an analytical standard and research reagent for biochemical and metabolic studies. It is supplied as a solid reference material for laboratory use only and is commonly used in analytical chemistry and metabolic profiling.
  • Provides a defined reference for analytical assays and method development.
  • High reported purity for accurate quantitation in research applications.
  • Suitable for metabolic and biochemical research studies.
  • Available in multiple small-scale pack sizes for bench-top experiments.
  • Stable solid form for convenient storage and handling.
Catalog No. 50-002-10205