Learn More
Medchemexpress LLC Imidazole-d4 | 6923-01-9 | 99.9% | 1 G
SDP
Supplier:  Medchemexpress LLC HYD0837S1G
Encompass_Preferred
Imidazole-d4 is a deuterium-labeled imidazole. This compound is commonly used as a tracer and an internal standard for quantitative analysis in various analytical techniques. Deuterium substitution has shown potential to influence the pharmacokinetic and metabolic profiles of drugs, making this compound valuable in drug development processes.
  • Deuterium labeled imidazole
  • Suitable as a tracer
  • Functions as an internal standard for quantitative analysis
  • Applicable in NMR, GC-MS, or LC-MS
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-10417