Learn More
Medchemexpress LLC Imidazole ketone era 100mg | 1801530-11-9 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY114481100MG
Encompass_Preferred
Imidazole ketone erastin is a small-molecule research reagent that potently and selectively inhibits the cystine-glutamate antiporter (system xc-) and induces ferroptosis in cellular models. It is used to study ferroptotic cell death mechanisms and evaluate anti-tumor effects in preclinical research.
  • Potent system xc- inhibitor and ferroptosis inducer.
  • High reported purity of 99.77%.
  • Available as solid and as a 10 mM solution in DMSO; common solid pack size is 100 mg.
  • Molecular formula C35H35ClN6O5, molecular weight 655.14 g·mol⁻¹.
  • Suitable for preclinical studies of ferroptosis and related tumor models.
Catalog No. 50-002-30073