Learn More
Cayman Chemical ImIdazole PropIonate 50mg
SDP
Supplier:  Cayman Chemical 3345850 MG
Encompass_Preferred
A metabolite of histidine; decreases protein levels of IRS1, IRS2, and phosphorylated Akt and increases levels of phosphorylated p70S6K in primary mouse hepatocytes; impairs glucose tolerance in germ-free and conventionally raised mice at 500 and 100 UG/animal, respectively; plasma levels are increased in patients with type 2 diabetes
Catalog No. 50-294-6505