Learn More
Medchemexpress LLC Imidodicarbonic acid 2-[6-nitro-3-(1H-pyrrolo[2,3-c]pyridin-1-yl)-5-isoquinolinyl] ester | 1841444-11-8 | 98.0% | 505.52 | C26H27N5O6 | 10MM 1ML
SDP
Supplier:  Medchemexpress LLC HY12622610MM 1ML
Encompass_Preferred
MK-6240 Precursor (CAS 1841444-11-8) is the synthetic precursor used to prepare [18F]-MK-6240, a tau-targeting positron emission tomography (PET) tracer for research applications. The material is provided as a powder (light yellow to yellow) and is also available as a prepared 10 mM solution in DMSO (1 mL). Certificate of analysis reports a high purity and confirms molecular weight and formula information; store the material per recommended conditions.
Catalog No. 50-002-39251