Learn More
Medchemexpress LLC Iminostilbene | 256-96-2 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYN7064
Encompass_Preferred
Iminostilbene, a chemical precursor of carbamazepine, is an orally active inhibitor of PKM2 (Pyruvate Kinase M2) and COX2 (Cyclooxygenase-2). It inhibits PKM2's interaction with HIF-1α and STAT3, reducing COX2 and iNOS expression. This suppresses macrophage-mediated inflammatory responses and improves myocardial ischemia/reperfusion (MI/R) injury, offering promise for research in inflammation regulation, cardiovascular, and macrophage-mediated immune-related diseases.
  • Precursor of carbamazepine
  • Orally active inhibitor of PKM2 and COX2
  • Inhibits PKM2 and its interaction with HIF-1α and STAT3
  • Reduces COX2 and iNOS expression
  • Suppresses LPS-induced inflammatory cytokine release
  • Suppresses macrophage-mediated inflammatory responses
  • Improves myocardial ischemia/reperfusion (MI/R) injury
  • Supports research in inflammation, cardiovascular, and immune-related diseases
Catalog No. 50-004-21567