Learn More
Medchemexpress LLC Imipenem | 64221-86-9 | 99.9% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB1369A
Encompass_Preferred
Imipenem (MK0787) is a stable crystalline derivative of thienamycin, functioning as an antibiotic with excellent activity against a broad spectrum of gram-positive and gram-negative aerobic and anaerobic bacteria. It is suitable for research on carbapenem-nonsusceptible and P. aeruginosa biofilm infections. This product is for research use only.
  • Excellent activity against a broad range of gram-positive and gram-negative aerobic and anaerobic bacteria.
  • Can be used for research on carbapenem-nonsusceptible and P. aeruginosa biofilm infections.
Catalog No. 50-004-14101