Learn More
AdipoGen Imiquimod (250 mg)
SDP
Supplier:  AdipoGen AGCR13569M250
Encompass_Preferred
Chemical. CAS: 99011-02-6. Formula: C14H16N4. MW: 240.3. Immune response modulator. Anti-inflammatory. TLR7 agonist. Stimulates proinflammatory cytokine production interferon-alpha (IFN-alpha), interleukin-6 (IL-6) and tumor necrosis factor-alpha (TNF-alpha) and activates NF-kappaB. Caspase-3 activator that directly induces procaspase-3 cleavage. Anti-cancer compound. Apoptosis inducer. Antiproliferative, independent of immune system activation or function. Antiviral and anti-angiogenic. Approved treatment for external genital warts caused by human papillomavirus infection.
Catalog No. 50-205-3938