Learn More
Medchemexpress LLC Imiquimod | 99011-02-6 | 100.0% | 200 MG
SDP
Supplier:  Medchemexpress LLC HYB0180200MG
Encompass_Preferred
Imiquimod is an immune response modifier that functions as a selective Toll-like Receptor 7 (TLR7) agonist. It demonstrates antiviral and antitumor effects in living organisms.
  • Selective TLR7 agonist.
  • Exhibits antiviral and antitumor properties in vivo.
  • Used for research related to external genital and perianal warts, cancer, and COVID-19.
  • Purity: 99.96%.
Catalog No. 50-004-06291