Learn More
Apexbio Technology LLC Imiquimod hydrochloride 99011-78-6 100mg
SDP
Supplier:  Apexbio Technology LLC B1186100
Encompass_Preferred
Imiquimod hydrochloride (CAS 99011-78-6) is a small-molecule agonist targeting Toll-like receptor 7 (TLR7) It is designed to activate TLR7 thereby triggering immune signaling cascades and promoting innate immune responses Imiquimod hydrochloride exerts its biological activity primarily through TLR7-mediated induction of pro-inflammatory cytokines such as IFN- TNF- and interleukin-12 In in vitro studies Imiquimod hydrochloride demonstrates TLR7 activation with an IC50 value of approximately 2 5 M varying according to experimental systems and conditions Based on these pharmacological properties Imiquimod hydrochloride holds research potential in studies of viral infections tumor immunotherapy and modulation of dermal immune responses
Catalog No. 50-000-92372