Learn More
Medchemexpress LLC Imiquimod (hydrochloride) | 99011-78-6 | 99.8% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0180A500MG
Encompass_Preferred
Imiquimod hydrochloride is an immune response modifier and a selective toll-like receptor 7 (TLR7) agonist. It demonstrates antiviral and antitumor effects in vivo and is used in research for various conditions, including external genital and perianal warts, cancer, and COVID-19.
  • Immune response modifier and selective TLR7 agonist
  • Demonstrates antiviral and antitumor effects
  • Used in research for external genital, perianal warts, cancer, and COVID-19
  • Soluble in DMSO and water
  • Can be utilized to induce psoriasis models
Catalog No. 50-004-07341