Learn More
TARGETMOL CHEMICALS INC Imirestat 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T155685MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes.Imirestat (HOE 843) is an aldose reductase inhibitor that can be used in diabetes research. Purity 99.08%
Catalog No. 50-287-8472