Learn More
Medchemexpress LLC Imisopasem manganese | 218791-21-0 | 98.2% | C21H31Cl2MnN5 | 10MG
SDP
Supplier:  Medchemexpress LLC HY1333610MG
Encompass_Preferred
Imisopasem manganese is a small-molecule, non-peptidyl mimetic of manganese superoxide dismutase (MnSOD) that functions as a superoxide dismutase mimetic. It removes superoxide anions while sparing other reactive species implicated in inflammatory responses, and is used as a research reagent in studies of oxidative stress, inflammation, and related signaling pathways.
  • Mimics MnSOD activity to scavenge superoxide anions.
  • Does not interfere with nitric oxide or peroxynitrite signaling.
  • Stable, low molecular weight synthetic compound.
  • Well characterized with a defined molecular formula and molecular weight.
  • Available in multiple solid pack sizes for research use.
Catalog No. 50-002-59036