Learn More
Medchemexpress LLC HY-13336 5mg Medchemexpress, Imisopasem manganese CAS:218791-21-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY133365MG
Encompass_Preferred
Medchemexpress, HY-13336 5mg Imisopasem manganese CAS:218791-21-0 Imisopasem manganese (M40403) is a stable non-peptidyl mimetic of manganese superoxide MnSOD. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1338