Learn More
Medchemexpress LLC Imlunestrant | 2408840-26-4 | 100.0% | C29H24F4N2O3 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY145572
Encompass_Preferred
Imlunestrant is an orally active, potent, and selective estrogen receptor degrader (SERD) with pure antagonistic properties. It leads to sustained inhibition of ER-dependent gene transcription and cell growth, making it suitable for research on ER-positive (ER+) advanced breast cancer (aBC) and endometrial endometrioid cancer (EEC).
  • Orally active, potent, and selective estrogen receptor degrader (SERD)
  • Pure antagonistic properties
  • Sustained inhibition of ER-dependent gene transcription and cell growth
  • Antiproliferative activity against human MCF7 cells harboring ERalpha Y537N mutant
  • Antiproliferative activity against human MCF7 cells harboring wild type ERalpha
  • Favorable pharmacokinetic (PK) properties
  • Demonstrates antitumor activity in ESR1 mutants
Catalog No. 50-003-57361