Learn More
Medchemexpress LLC Immepip dihydrobromide | 164391-47-3 | MFCD06407876 | 99.9% | 327.06 | C9H17Br2N3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY101321A50MG
Encompass_Preferred
Immepip dihydrobromide is the dihydrobromide salt of immepip, a potent histamine H3 receptor agonist used in neuroscience research to probe H3-mediated modulation of histaminergic signaling. It has been reported to reduce cortical histamine release and is used to study neurological disease mechanisms.
  • Potent histamine H3 receptor agonist suitable for receptor pharmacology studies.
  • Reduces cortical histamine release in reported studies.
  • High purity for research use (listed as 99.89%).
  • Supplied as a crystalline dihydrobromide salt for experimental consistency.
  • Available in multiple pack sizes, including 50 mg, for laboratory flexibility.
Catalog No. 50-002-11214