Learn More
Electron Microscopy Sciences Cargille Immersion Oil Type LDF 1 OZ
SDP
Supplier:  Electron Microscopy Sciences 16919
Encompass_Preferred
After years of development and field testing, we now have available two new PCB-Free Immersion Oils for Fluorescence Microscopy. Extremely low fluorescence is achieved by Type LDF. Possesses the highest resolution of the oils for fluorescence applications. Replacement for type DF.
Catalog No. 50-365-631