Learn More
ACCEGEN BIOTECH Immortalized Human Gastric Fibroblasts (hTert)
Supplier:  ACCEGEN BIOTECH ABIH0008X
Encompass
Human Gastric Fibroblasts (HGFs) are initiated by elutriation from dissociated human gastric tissue. Puromycin resistant hTert-HGFs are selected from HGFs after infected with lentiviruses expressing hTert. The cells are shipped in frozen vials.
Catalog No. NC2749838