Learn More
Cusabio Technology LLC Human Immunoglobulin E,IgE ELISA Kit
Supplier:  Cusabio Technology LLC CSBE04980H
Encompass_Preferred
This Human IgE ELISA Kit was designed for the quantitative measurement of Human IgE protein in serum, plasma. It is a Sandwich ELISA kit, its detection range is 62.5 ng/mL-4000 ng/mL and the sensitivity is 3.640 ng/mL. One kit contains 96 wells.
Catalog No. NC2952803