Learn More
Electron Microscopy Sciences Aurion ImmunoGold EM Pra-6/Pra-10/BSA-C Kit
SDP
Supplier:  Electron Microscopy Sciences 25548
Encompass_Preferred
Immunogold reagent kit. Two non-overlapping gold particle sizes, Coupled to either Goat-anti-Rabbit and Goat-anti-Mouse IgG/IgM antibodies, includes: 1ml Protein A 6 nm, 1ml Protein A 10 nm, 30ml BSA-c background suppressing agent.
Catalog No. 50-282-87